Learn More
STEMCELL Technologies Alpha MEM without Nucleosides
Artikelnummer. 17168091
Encompass
Click to view available options
Menge:
500 ml
Packungsgröße:
500 Milliliter
Artikelnummer. 17168091
Marke: STEMCELL Technologies 36453
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Minimum Essential Medium Eagle - alpha modification (Alpha MEM) without nucleosides
  • Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications
  • Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition
  • MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium
Spezifikation
Menge 500 ml