Learn More
Bax Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610983
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Proper development and maintenance of homeostasis within tissues is achieved through cell proliferation, differentiation, and apoptosis. Members of the Bcl-2 protein family function to either inhibit (Bcl-2 and Bcl-XL) or promote (Bax and Bak) apoptosis.
Immunofluorescence, Immunoprecipitation, Western Blotting
Spezifikation
Bax | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Bax-α aa. 55-178 | |
150 μg | |
Cell Biology | |
Canine, Human | |
IgG1 |
Western Blot | |
3 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.