Learn More
CD11A Mouse anti-Human, Brilliant Violet 510, Clone: HI111, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 563480
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).
Spezifikation
CD11a | |
Monoclonal | |
Brilliant Violet 510 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
HI111 | |
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.