Learn More
CD14 Mouse anti-Human, Brilliant Violet 711, Clone: MPHIP9, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 563372
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The M φP9 monoclonal antibody specifically binds to human CD14, a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, this CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Spezifikation
CD14 | |
Monoclonal | |
Brilliant Violet 711 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Monocytes | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MPHIP9 | |
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
For Research Use Only.