Learn More
CD161a Mouse anti-Rat, FITC, Clone: 10/78, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555008
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The 10/78 antibody reacts with NKR-P1A, a 60-kDa homodimer expressed on all natural killer (NK) cells and a small subset of T lymphocytes. The 10/78 antibody competes with the previously described 3.2.3 antibody for binding to the antigen. NKR-P1A is a type-II integral membrane protein with an extracellular C-type lectin domain, which is an NK cell-activating receptor specific for tumor target cells. Many rat dendritic cells have been shown to express NKR-P1A, and a subpopulation of these cells has cytotoxic activity. NKR-P1A has also been detected at low levels on peripheral blood monocytes, and its expression is upregulated in IFN-γ-activated monocytes, specifically in a subpopulation of large monocytes with phagocytic capacity. Furthermore, activated peripheral blood neutrophils may express a low level of NKR-P1A. In the mouse and rat, three members of the NKR-P1 gene family have been identified; but in the human gene family, a single NKR-P1 homologue has been discovered and designated Cd161
Spezifikation
CD161a | |
Monoclonal | |
0.5mg/mL | |
NKR-P1A | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
Oct-78 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not reported | |
0.5 mg | |
Immunology | |
Rat | |
IgG1 κ |
For Research Use Only.