Learn More
CD209 Mouse anti-Human, PE, Clone: DCN46, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561765
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
The DCN46 antibody specifically binds to dendritic cell-specific ICAM-3 grabbing nonintegrin (DC-SIGN or CD209), a type-II membrane protein of approximately 44 kDa with a mannose-binding C-type lectin domain. It is highly expressed on dendritic cells in mucosal tissues. Its sequence is identical to the HIV-1 envelope gp120-binding C-type lectin, and reports suggest that DC-SIGN binds to HIV-1 gp120 and effectively transmits infectious HIV-1 to resting T lymphocytes expressing CD4 and chemokine receptors. The C-type lectin domain of DC-SIGN is also capable of binding other pathogenic viruses, bacteria, and parasites. Reports also suggest that DC-SIGN enables the highly efficient migration of dendritic cells from blood into the tissues. It can interact with ICAM-2, which has a similar sequence as ICAM-3, and is abundantly expressed on vascular and lymphoid endothelium. Thus, DC-SIGN mediates dendritic cells rolling and transendothelial migration, and its interaction with ICAM-2 is essential to specific migratory functions of dendritic cells.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD209 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Monocyte Derived DC Cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
DCN46 | |
DC-SIGN | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
For Research Use Only.