Learn More
CD80 Hamster anti-Mouse, FITC, Clone: 16-10A1, BD
Hamster Monoclonal Antibody
Marke: BD Biosciences 561954
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with FITC under optimum conditions, and unreacted FITC was removed. CD80 has been constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages and it is inducible on B cells by various means.
Host Species: Hamster
Clone: 16-10A1
Isotype: IgG2, κ
Species Reactivity: Mouse
Immunogen: Mouse CD80 (B7) Transfected Cell Line
- Activation of purified B cells with LPS can induce CD80 expression in as few as 18 hour
- 16-10A1 antibody blocks binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines
Flow Cytometry
Spezifikation
CD80 | |
Monoclonal | |
0.5mg/mL | |
Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1 | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 κ |
Flow Cytometry | |
16-10A1 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CD80 (B7) Transfected Cell Line | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.