missing translation for 'onlineSavingsMsg'
Learn More

CD8a Rat anti-Mouse, PerCP, Clone: 53-6.7, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 561092

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15890759

  • 171.00 € / 1 Stück
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

The 53-6.7 antibody monoclonal antibody specifically binds to the 38 kDa α and 34 kDa α chains of the CD8 differentiation antigen (Ly-2 or Lyt-2) of all mouse strains tested. The CD8 α and α chains (CD8a) form heterodimers with the CD8 β chain (CD8b, Ly-3, or Lyt-3) on the surface of most thymocytes. A subpopulation of mature T lymphocytes (i.e., MHC class I-restricted T cells, including most T suppressor/cytotoxic cells) expresses almost exclusively the CD8 αβ heterodimer (the αα chains arise from alternatively spliced messengers of a single CD8a gene. The longer α form associates with p56 [lck] via a CXCP motif in its cytoplasmic domain, which it shares with CD4, but not with CD8b. The truncated α chain is unable to associate with p56 [lck], and it may function to attenuate the CD8-mediated costimulatory signal during intrathymic T-cell maturation. In vivo and in vitro treatment with 53-6.7 mAb has reportedly been effective at depleting CD8+ peripheral T lymphocytes. The 53-6.7 antibody has also been reported to cross-reaact with CD8 α- and α-like polypeptides on subsets of thymic and peripheral lymphocytes in the Egyptian toad, Bufo regularis.

Specifications

Specifications

CD8a
Monoclonal
0.2mg/mL
Cd8a; CD8 alpha chain; Ly-2; Lyt2; Lyt-2; Ly-35; Ly-B
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
53-6.7
PerCP
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Spleen Cells or Thymocyte Membranes
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents
Product Certifications
Special Offers

Special Offers

For Research Use Only.