missing translation for 'onlineSavingsMsg'
Learn More

CD95 Mouse anti-Human, PE-Cy5, Clone: DX2, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 559773

Weitere Details : Netto-Gewicht : 0.02000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15879768

  • 415.00 € / 1 Stück
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Reacts with the 45kDa transmembrane cell surface Fas or Apo-1 antigen (designated CD95 at the Fifth HLDA Workshop) which is expressed on a variety of normal and neoplastic cells. The Fas/Apo-1 antigen is a polypeptide, which plays a role in the programmed sequence of events leading to cell death, termed apoptosis. The DX2 clone specifically reacts with murine L cells, murine L1210 leukemia cells and murine P815 mastocytoma cells transfected with human Fas cDNA but not with untransfected parental cell lines. Cross-linking with DX2 delivers an apoptotic signal indicating that DX2 recognizes a functional epitope of the CD95 antigen. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

Spezifikation

Spezifikation

CD95
Monoclonal
PE-Cyanine5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD95-transfected L Cells
100 Tests
Immunology
Baboon, Canine, Cynomolgus, Human, Porcine, Rhesus Monkey
IgG1 κ
Flow Cytometry
DX2
APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.