Learn More
CTCF Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 612148
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The family of nucleic acid-binding C2H2 type zinc finger transcription factors is divided into two classes. One class consists of small proteins (Gli1, Krox-20, WT1, Egr-1, and Sp1) with conserved zinc finger clusters of 3 to 5 units, while the other class (ZNF91, ZNF74, ZFP37, CTCF) can contain more than 10 zinc finger clusters. CTCF is a ubiquitously expressed, highly conserved transcription factor that contains 10 C2H2- and 1 C2H-type zinc-finger motifs. CTCF binds to and represses transcription at the promoter-proximal regions of c-myc oncogenes,
while CTCF can bind to and activate transcription at the promoter for amyloid β. Another mechanism of CTCF-mediated repression may include binding to insulator regions between enhancers and promoters resulting in enhancer blocking. In addition, CTCF gene may be a candidate tumor suppressor gene, since it localizes to a narrow cancer-associated chromosome region, and has been shown to have tumor-specific rearrangements in breast cancer patients. Thus, CTCF transcriptional regulation may be important for cell cycle progression, differentiation, apoptosis, and tumorigenesis.
Immunofluorescence, Western Blotting
Spezifikation
CTCF | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human CTCF aa. 184-290 | |
50 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
48 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.