Learn More
IL-5 Rat anti-Mouse/Human, Unlabeled, Clone: TRFK5, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554393
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and has been reported to cross-react with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Host Species: Rat
Clone: TRFK5
Isotype: IgG1 κ
Species Reactivity: Mouse, Human
Immunogen: Mouse Semi-Purified T-Cell Clone Supernatant
Functional Assay, Neutralization
Spezifikation
IL-5 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Semi-Purified T-Cell Clone Supernatant | |
0.5 mg | |
Immunology | |
Human, Mouse | |
IgG1 κ |
ELISA | |
TRFK5 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.