missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase Cg (pY783) Mouse, Unlabeled, Clone: 27, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 612465

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15896189

  • 696.54 € / 1 Stück
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol phosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLCγ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with receptor tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: 1) PLCγ1, which is ubiquitously expressed, and 2) PLCγ2, found primarily in the lymphoid system. PLCγ is essential for growth factor-induced cell motility and mitogenesis. PLCγ1-null mice exhibit retarded embryonic growth and lethality in midgestation. In addition, PDGF stimulation leads to phosphorylation of PLCγ at Tyr 783, and activation of hydrolyzing activity. Overexpression of PLCγ is evident in several forms of cancer and it has been identified as a key mediator of PDGF-dependent cellular transformation. Thus, regulation of PLCγ activity by growth factors is involved in cell growth and transformation.

Western Blotting

Spezifikation

Spezifikation

Phospholipase Cg (pY783)
Monoclonal
250μg/mL
PLCγ (pY783)
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
27
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Phosphorylated PLCγ1 Peptide
150 μg
Cell Biology
Rat, Mouse, Human
IgG1
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.