missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP50 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 27355-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The heterotetrameric coat assembly protein complex, also known as the adaptor-related protein complex 2 (AP-2), belongs to the adaptor complexes medium subunits family. The mu 1 subunit of the AP-2 complex (AP2M1) is required for the activity of a vacuolar ATPase, which is responsible for proton pumping occurring in the acidification of endosomes and lysosomes. AP2M1 has also been shown to associate with the HIV-1 protein Nef, suggesting that Nef may use AP-2 complex to enhance the rate of endocytosis of both CD4 and class I MHC. AP2M1 may also play an important role in regulating the intracellular trafficking and function of cytotoxic T-lymphocyte associated (CTLA)-4 protein. At least two isoforms of AP2M1 are known to exist.Spezifikation
AP50 | |
Polyclonal | |
Unconjugated | |
AP2M1 | |
Adaptin mu2, AP 2 complex subunit mu, AP 2 mu chain, AP2M1, AP50, CLAPM1, HA2 50 kDa subunit, KIAA0109, mu2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116563, 1173, 11773 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P84091, P84092, Q96CW1 | |
Ap2m1 | |
AP50 Fusion Protein Ag26059 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |