Learn More
STEMCELL Technologies ArciTect™ T7 Endonuclease I Kit
Artikelnummer. 17139311
Encompass_Preferred
Change view
Click to view available options
Menge:
125 Reaktionen
25 Reaktionen
Packungsgröße:
1 Set
Artikelnummer. Menge unitSize
17139311 125 Reaktionen -
17129311 25 Reaktionen 1 Set
2 options
Artikelnummer. 17139311
Marke: STEMCELL Technologies 76022
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
For estimation of CRISPR-Cas9 genome editing efficiency
  • ArciTect™ T7 Endonuclease I is the preferred enzyme for detecting genome editing such as insertions or deletions (INDELs) generated by CRISPR-Cas9
  • ArciTect™ T7 Endonuclease I Kit is comprised of ArciTect™ T7 Endonuclease I and ArciTect™ T7 Endonuclease I Buffer (10X), which have been tested and validated for use with the ArciTect™ CRISPR-Cas9 genome editing system
  • ArciTect™ T7 Endonuclease I recognizes and cleaves mismatched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA, and, less efficiently, nicked double-stranded DNA
  • Since the cleavage efficiency is proportional to the number of INDELs created at a specific DNA target, ArciTect™ T7 Endonuclease I Kit is used to estimate gene-editing efficiency in a rapid and cost-effective manner
Specifica
Zur Verwendung mit (Anwendung) Estimation of CRISPR-Cas9 Genome Editing Efficiency
Menge 125 Reaktionen
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.