Learn More
STEMCELL Technologies ArciTect™ T7 Endonuclease I Kit
Click to view available options
Menge:
125 Reaktionen
25 Reaktionen
Packungsgröße:
1 Set
Beschreibung
- ArciTect™ T7 Endonuclease I is the preferred enzyme for detecting genome editing such as insertions or deletions (INDELs) generated by CRISPR-Cas9
- ArciTect™ T7 Endonuclease I Kit is comprised of ArciTect™ T7 Endonuclease I and ArciTect™ T7 Endonuclease I Buffer (10X), which have been tested and validated for use with the ArciTect™ CRISPR-Cas9 genome editing system
- ArciTect™ T7 Endonuclease I recognizes and cleaves mismatched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA, and, less efficiently, nicked double-stranded DNA
- Since the cleavage efficiency is proportional to the number of INDELs created at a specific DNA target, ArciTect™ T7 Endonuclease I Kit is used to estimate gene-editing efficiency in a rapid and cost-effective manner
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Estimation of CRISPR-Cas9 Genome Editing Efficiency |
| Menge | 25 Reaktionen |