missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14349-1-AP-20UL
Informations supplémentaires : Netto-Gewicht : 0.00010kg
Description
ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.Spécification
ARHGAP25 | |
Polyclonal | |
Unconjugated | |
ARHGAP25 | |
ARHGAP25, KAIA0053, KIAA0053 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
232201, 500246, 9938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42331, Q8BYW1 | |
Arhgap25 | |
ARHGAP25 Fusion Protein Ag5691 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |