Learn More
DiscoverX AssayComplete™ Cell Culture Kit-108
Click to view available options
Menge:
1 Kit
Beschreibung
AssayComplete™ Cell Culture Kit-108 is an optimized media kit intended for use in culturing and propogating DiscoverX cell lines. This kit contains the base medium, relevant serum, and pen/strep required to produce a full cell culture medium for optimal cell culture results. This product is provided as a 3-component kit which will produce approximately 550mL of finished media. Please refer to cell line specific datasheet to determine the recommended cell culture kit.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Cell Culture Kits |
| Menge | 1 Kit |