Learn More
DiscoverX AssayComplete™ Cell Culture Kit-113
Artikelnummer. 17332659
Encompass
Click to view available options
Menge:
1 Kit
Artikelnummer. 17332659
Marke: DiscoverX 923113G
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
AssayComplete™ Cell Culture Kit-113 is a media kit including base medium, serum and pen/strep for use in culturing DiscoverX cell lines. This product is provided as a 3-component kit which produces ≈550mL of media.
AssayComplete™ Cell Culture Kit-113 is an optimized media kit intended for use in culturing and propogating DiscoverX cell lines. This kit contains the base medium, relevant serum, and pen/strep required to produce a full cell culture medium for optimal cell culture results. This product is provided as a 3-component kit which will produce approximately 550mL of finished media. Please refer to cell line specific datasheet to determine the recommended cell culture kit.
Spezifikation
Zur Verwendung mit (Anwendung) Cell Culture Kits
Menge 1 Kit