missing translation for 'onlineSavingsMsg'
Learn More

BCL2L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 16026-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16885474

  • 329.85 € / 150 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The "rox" portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.
Spezifikation

Spezifikation

BCL2L2
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70345, Q92843
Apoptosis regulator Bcl W, BCL2L2, KIAA0271
Rabbit
IgG
150 μL
-20°C
Primary
12050, 599, 60434
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BCL2L2
Liquid
BCL2L2
BCL2L2 Fusion Protein Ag8863
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation