missing translation for 'onlineSavingsMsg'
Learn More

Biochanin A, MedChemExpress

Artikelnummer. 16421136
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
200 mg
500 mg
Packungsgröße:
500 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
16421136 200 mg -
16461736 500 mg 500 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16421136 Lieferant Medchem Express Lieferanten-Nr. HY14595
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Biochanin A is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, which inhibits FAAH with IC50s of 1.8, 1.4 and 2.4 μM for mouse, rat, and human FAAH, respectively.

Biochanin A is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, which inhibits FAAH with IC50s of 1.8, 1.4 and 2.4 μM for mouse, rat, and human FAAH, respectively.

Specifications

Chemischer Name oder Material Biochanin A
CAS 491-80-5
Farbe Light Yellow
Hinweise zur Reinheitsqualität Research
Summenformel C16H12O5
Menge 200 mg
Synonym 4-Methylgenistein Olmelin
Löslichkeitsinformationen DMSO : ≥ 50 mg/mL (175.90 mM)
SMILES OC1=C2C(OC=C(C3=CC=C(OC)C=C3)C2=O)=CC(O)=C1
Molekulargewicht (g/mol) 284.26
Formelmasse 284.26
Reinheit (%) 98.62%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.