Learn More
ACROBiosystems Biotinylated Human M-CSF / CSF-1 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems MCF-H82E6-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The colony stimulating factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF), is a secreted cytokine which influences hematopoietic stem cells to differentiate into macrophages or other related cell types. Eukaryotic cells also produce M-CSF in order to combat intercellular viral infection. It is one of the three experimentally described colony-stimulating factors. M-CSF binds to the colony stimulating factor 1 receptor. Macrophage colony-stimulating factor has been shown to interact with PIK3R2. M-CSF (or CSF-1) is a hematopoietic growth factor that is involved in the proliferation, differentiation, and survival of monocytes, macrophages, and bone marrow progenitor cells. Locally produced M-CSF in the vessel wall contributes to the development and progression of atherosclerosis.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.8 kDa. The protein migrates as 35-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_757349.1 | |
25 μg | |
Glu 33 - Arg 255 | |
M-CSF,CSF-1,Lanimostim | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
28.8 kDa | |
Biotinylated Human M-CSF, His,Avitag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Accession # P09603-1). | |
-20°C | |
M-CSF | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |