Learn More
ACROBiosystems Biotinylated Human M-CSF / CSF-1 Protein, His, Avitag™
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_757349.1 |
---|---|
Molekulargewicht | 28.8 kDa |
Quelle | Biotinylated Human M-CSF, His,Avitag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Accession # P09603-1). |
Immunogen | Glu 33 - Arg 255 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16417869
|
ACROBiosystems
MCF-H82E6-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16427869
|
ACROBiosystems
MCF-H82E6-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The colony stimulating factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF), is a secreted cytokine which influences hematopoietic stem cells to differentiate into macrophages or other related cell types. Eukaryotic cells also produce M-CSF in order to combat intercellular viral infection. It is one of the three experimentally described colony-stimulating factors. M-CSF binds to the colony stimulating factor 1 receptor. Macrophage colony-stimulating factor has been shown to interact with PIK3R2. M-CSF (or CSF-1) is a hematopoietic growth factor that is involved in the proliferation, differentiation, and survival of monocytes, macrophages, and bone marrow progenitor cells. Locally produced M-CSF in the vessel wall contributes to the development and progression of atherosclerosis.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.8 kDa. The protein migrates as 35-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_757349.1 | |
Biotinylated Human M-CSF, His,Avitag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Accession # P09603-1). | |
-20°C | |
M-CSF | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
28.8 kDa | |
Glu 33 - Arg 255 | |
M-CSF,CSF-1,Lanimostim | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |