Learn More
ACROBiosystems Biotinylated Human Mesothelin / MSLN (296-580) Protein, His,Avitag™
MALS verified
Marke: ACROBiosystems MSN-H82E9-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 35.9 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH09272.1 | |
200 μg | |
Glu 296 - Gly 580 | |
MSLN,Mesothelin,MPF | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
35.9 kDa | |
Biotinylated Human Mesothelin (296-580), His,Avitag (MSN-H82E9) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). | |
-20°C | |
Mesothelin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |