Learn More
ACROBiosystems Biotinylated Human Siglec-2 / CD22 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems SI2-H82F8-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
B-cell receptor CD22 is also known as Sialic acid-binding Ig-like lectin 2 (Siglec-2), B-lymphocyte cell adhesion molecule (BL-CAM), T-cell surface antigen Leu-14, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. CD22 mediates B-cell B-cell interactions, and may be involved in the localization of B-cells in lymphoid tissues. Siglec-2 / CD22 binds sialylated glycoproteins, one of which is CD45. Siglec2 / CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 103.9 kDa. The protein migrates as 115-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001762.2 | |
200 μg | |
Asp 20 - Arg 687 | |
CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,SIGLEC2FLJ22814 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
103.9 kDa | |
Biotinylated Human Siglec-2, Fc,Avitag (SI2-H82F8) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1). | |
-20°C | |
Siglec-2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |