Learn More
ACROBiosystems Biotinylated Mouse CD27 Ligand / CD70 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems CDL-M82Qb-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Cluster of Differentiation 70 (CD70) is also known as CD27 ligand (CD27L / CD27LG), TNFSF7, TNFSF7G, is a type II transmembrane glycoprotein belonging to the TNF superfamily (TNFSF) and is a surface antigen found on activated T-and B-lymphocytes and mature dendritic cells. Binding of CD70 to its receptor CD27 induces in priming, effector functions, differentiation and memory formation of T-cells, and thus is invloved in the biological processes including T-cell activation, the proliferation of costimulated T-cells, as well as the generation of cytolytic T-cells. CD70 on T cells provides costimulatory signals that are required for T cell proliferation, clonal expansion and the promotion of effector T cell formation. CD70 on mouse B cell has been shown to inhibit terminal differentiation of activated B cells into plasma cells and enhances commitment to memory B cell responses. CD70 induces proliferation and IFNγ production, on NK cells.
This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.1 kDa. The protein migrates as 28-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_035747.1 | |
200 μg | |
Gln 47 - Pro 195 | |
CD70,CD27LG,TNFSF7,TNFSF7G,CD27L | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
20.1 kDa | |
Biotinylated Mouse CD27 Ligand, His,Avitag (CDL-M82Qb) is expressed from human 293 cells (HEK293). It contains AA Gln 47 - Pro 195 (Accession # O55237-1). | |
-20°C | |
CD27 Ligand | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |