Learn More
Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein
Human BMPR1B (ALK6), GST Tag Recombinant Protein
Marke: Invitrogen™ PV6236
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
BMPR1B is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Spezifikation
66.7 kDa | |
ALK-6, ALK6, CDw293 | |
Human | |
GST Tag | |
Serine/Threonine Kinase | |
Recombinant |
100μg | |
aa 149-end | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Liquid |