missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BRAF, GST Tag Recombinant Protein
BRAF Recombinant Human Protein
Marke: Invitrogen PV3848
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron.Spezifikation
NP_004324.2 | |
2353, 673 | |
10 μg | |
RAF Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
v-raf murine sarcoma viral oncogene homolog B1 | |
Kinases & Inhibitors | |
Liquid | |
Oncology, Signal Transduction |
Kinase Assay | |
113.6 kDa | |
BRAF, BRAF1 | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
BRAF | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |