missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf93 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25364-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
C16orf93, also named as CCDC189, is a coiled-coil domain-containing protein which has four isoforms with MW 38 kDa, 34 kDa, 12 kDa and 20 kDa.Spezifikation
C16orf93 | |
Polyclonal | |
Unconjugated | |
CCDC189 | |
C16orf93, CCDC189 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90835 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1A4V9 | |
CCDC189 | |
C16orf93 Fusion Protein Ag21676 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |