missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25953-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
C20orf3 may play a role in adipocyte differentiation.Spezifikation
C20orf3 | |
Polyclonal | |
Unconjugated | |
APMAP | |
APMAP, BSCv, C20orf3, Protein BSCv | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57136 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HDC9 | |
APMAP | |
C20orf3 Fusion Protein Ag23029 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |