missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CAMK4 (CaMKIV), GST Tag Recombinant Protein
Human CAMK4 (CaMKIV), GST Tag Recombinant Protein
Marke: Thermo Scientific™ PV3979
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
CAMK4 (CaMKIV) has limited tissue distribution, and has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.Spezifikation
NP_001735 | |
79.6 kDa | |
CAMK4 (CaMKIV) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CAMK Group | |
E. coli | |
Frozen |
Kinase Assay | |
1 mg | |
CAMK1 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Full Length | |
Drug Development |