missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Rat anti-Mouse,Human, BB700, Clone: M1/70, BD Horizon™
Rat Monoclonal Antibody
Marke: BD Biosciences 566417
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
CD11b Monoclonal antibody specifically detects CD11b in Mouse,Human samples. It is validated for Flow Cytometry.Spezifikation
CD11b | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse, Human | |
Purified |
Flow Cytometry | |
M1/70 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |