Learn More
CD13 Mouse anti-Human, PE, Clone: WM15, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560998
Additional Details : Netto-Gewicht : 0.01000kg
Description
The WM15 monoclonal antibody specifically binds to the 150 kDa Type II integral membrane glycoprotein aminopeptidase N. This antibody binds to GM-progenitor cells, granulocytic and monocytic cells, and mast cells, but not to lymphocytes, platelets or erythrocytes. Aminopeptidase N is involved in the metabolism of many regulatory peptides.
Specifications
CD13 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
WM15 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human AML Cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.