missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD184 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: 12G5, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566553
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD184 Monoclonal antibody specifically detects CD184 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Spezifikation
CD184 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Flow Cytometry | |
12G5 | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Purified |