missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, V450, Clone: HIB19, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB30-V450-25Tests
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CD19 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD19 | |
Protein A/G Purified | |
RUO | |
930 | |
IgG1 κ |
Flow Cytometry | |
HIB19 | |
Violet Fluor 450 (V450) | |
NP_001171569 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |