Learn More
CD2 Rat, FITC, Clone: RM2-5, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB89-F-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CD2 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd2 | |
Protein A/G Purified | |
RUO | |
12481 | |
IgG2b λ |
Flow Cytometry | |
RM2-5 | |
FITC | |
NP_038514 | |
Rat | |
500 μg | |
Primary | |
Store at 2-4°C. |