missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFLAR/FLIP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10394-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.Spezifikation
CFLAR/FLIP | |
Polyclonal | |
Unconjugated | |
CFLAR | |
c FLIP, c FLIPL, c FLIPR, c FLIPS, CASH, CASP8AP1, Caspase eight related protein, Caspase homolog, Casper, CFLAR, CLARP, FLAME, FLAME 1, FLAME1, FLIP, I FLICE, Inhibitor of FLICE, MRIT, Usurpin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8837 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence | |
0.72 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15519 | |
CFLAR | |
CFLAR/FLIP Fusion Protein Ag0593 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |