Learn More
STEMCELL Technologies Collagenase A, ACF
Artikelnummer. 17179261
Encompass_Preferred
Click to view available options
Menge:
100 mg
1 g
Packungsgröße:
1 Gramm
Artikelnummer. 17179261
Marke: STEMCELL Technologies 07435
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Animal component-free collagenase for the digestion of native collagen fibrils
  • Collagenase A, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
  • Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
  • Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
  • Collagenase A, ACF contains levels of proteolytic activity similar to type I and type II collagenases
  • Collagenase A, ACF is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Spezifikation
Zur Verwendung mit (Anwendung) Digestion of Native Collagen Fibrils
Menge 1 g