Learn More
STEMCELL Technologies Collagenase A, ACF
Click to view available options
Menge:
100 mg
1 g
Packungsgröße:
1 Gramm
Beschreibung
- Collagenase A, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
- Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
- Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
- Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
- Collagenase A, ACF contains levels of proteolytic activity similar to type I and type II collagenases
- Collagenase A, ACF is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Digestion of Native Collagen Fibrils |
| Menge | 100 mg |