Learn More
BUCHI Condensers for BUCHI Rotavapors
Alle ansehen Buchi Produkte
Click to view available options
Typ:
Kondensator S
Vertikalkondensator V
Material (Beschichtung):
P+G
Unbeschichtet
Unit Size:
1 Stück
Description
- High throughput due to advanced design of condensers
- Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
- Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
- Maximum stability and safety with plastic and glass coating
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Typ | Vertikalkondensator V |
| Zur Verwendung mit (Geräte) | Rotavapor R-210/R-215 |
| Material (Beschichtung) | P+G |