missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11709-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.Spezifikation
CSN1 | |
Polyclonal | |
Unconjugated | |
GPS1 | |
COPS1, CSN1, G protein pathway suppressor 1, GPS 1, GPS1, Protein MFH, SGN1, Signalosome subunit 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2873 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13098 | |
GPS1 | |
CSN1 Fusion Protein Ag2273 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |