missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10721-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Cysteine and glycine-rich protein 3 (CSRP3) is a member of the CSRP family of genes encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in CRP3 is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Very strongly similar to rat MLP; contains zinc-finger LIM domains. Other genes in the family include CSRP1 and CSRP2.Spezifikation
CSRP3 | |
Polyclonal | |
Unconjugated | |
CSRP3 | |
Cardiac LIM protein, CLP, CMD1M, CMH12, CRP3, CSRP3, Cysteine rich protein 3, LIM domain protein, cardiac, LMO4, MLP, Muscle LIM protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117505, 13009, 8048 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50461, P50462, P50463 | |
CSRP3 | |
CSRP3 Fusion Protein Ag1136 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |