missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 51030-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. CTRP6 contains at least 4 glycosylation motifs, suggesting that CTRP6 may be highly post-translationally modified.Spezifikation
CTRP6 | |
Polyclonal | |
Unconjugated | |
C1QTNF6 | |
C1QTNF6, CTRP6, ZACRP6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114904 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BXI9 | |
C1QTNF6 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |