Learn More
ACROBiosystems Cynomolgus CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Flag Tag
MALS verified
Marke: ACROBiosystems CDD-C52W4-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Cynomolgus CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag (MALS verified) is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with his tag at the C-terminus and subunit CD3D isSpezifikation
15.5 kDa (CD3E) and 14.5 kDa (CD3D) | |
Cynomolgus CD3E&CD3D; Heterodimer Protein, His Tag&Flag; Tag (CDD-C52W4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5 (CD3E) and Q95LI8 (CD3D)). | |
-20°C | |
CD3E and CD3D | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
1 mg | |
Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
His Tag and Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |