missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Flag Tag
MALS verified
447.57 € - 2794.30 €
Spezifikation
Molekulargewicht | 15.5 kDa (CD3E) and 14.5 kDa (CD3D) |
---|---|
Quelle | Cynomolgus CD3E&CD3D; Heterodimer Protein, His Tag&Flag; Tag (CDD-C52W4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5 (CD3E) and Q95LI8 (CD3D)). |
Immunogen | Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) |
Lagerungsbedingungen | -20°C |
Gen-Alias | CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15846377
|
ACROBiosystems
CDD-C52W4-100UG |
100 μg |
447.57 €
100 Mikrogramm |
Verfügbar ab: 31-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15856377
|
ACROBiosystems
CDD-C52W4-1MG |
1 mg |
2794.30 €
1 Milligramm |
Verfügbar ab: 31-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Cynomolgus CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag (MALS verified) is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with his tag at the C-terminus and subunit CD3D isSpezifikation
15.5 kDa (CD3E) and 14.5 kDa (CD3D) | |
Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
His Tag and Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Cynomolgus CD3E&CD3D; Heterodimer Protein, His Tag&Flag; Tag (CDD-C52W4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5 (CD3E) and Q95LI8 (CD3D)). | |
-20°C | |
CD3E and CD3D | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |