Learn More
ACROBiosystems Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free
MALS verified
Marke: ACROBiosystems CDG-C52W6-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.0 kDa (CD3E) and 14.4 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G containSpezifikation
NP_001270544.1 (CD3E) and NP_001270839.1 (CD3G) | |
1 mg | |
Gln 22 - Asp 117 (CD3E) and Gln 23 - Thr 113 (CD3G) | |
CD3 epsilon and CD3 gamma,CD3E and CD3G | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
16.0 kDa (CD3E) and 14.4 kDa (CD3G) | |
Cynomolgus CD3E&CD3G; Heterodimer Protein, His Tag&Tag; Free (CDG-C52W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) and Q95LI7-1 (CD3G)). | |
-20°C | |
CD3E and CD3G | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |