Learn More
ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, Fc Tag
Marke:  ACROBiosystems GP3-C5256-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15886867
  • 2258.12 € / 1 Milligramm
Verfügbar ab: 11-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.4 kDa respectively
Spezifikation
Spezifikation
XP_005594665.1
38.1 kDa and 49.4 kDa
1 mg
Gln 25 - His 559
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Cynomolgus
Powder
>90% as determined by SDS-PAGE
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).
-20°C
Glypican 3
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens