Learn More
Biomex™ Cytomegalovirus (CMV) Seroconversion Panels
Biomex™ Cytomegalovirus (CMV) Seroconversion Panels are intended for standard testing by diagnostic manufacturers and researchers during assay development, evaluation, troubleshooting and post-marked surveillance of IgM/IgG antibody test systems and methods.
Marke: Biomex™ SCP-CMV-006
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Illustrates the onset and decline of IgM and IgG Cytomegalovirus antibodies from one individual over a period of time.
- For CMV assay evaluation purposes of IgM/IgG antibody test systems.
- Serves as validation tool for diagnostic sensitivity, determination of analytical sensitivity, identification of cut-off values or to study the humoral immune response to this infection.
- Defibrinated and filtered.
- Sodium azide (0.095%) was added as preservative.
- All panel members are ready to use.
Spezifikation
Single donor | |
24 x 0.5 mL Vials | |
−20°C to -80°C | |
Cytomegalovirus |