missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosinethylester Hydrochlorid, 98 %, Thermo Scientific™

Catalog Number p-7041464
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
23234-43-7
C11H16ClNO3
245.70
MFCD00180750
BQULAXAVRFIAHN-OXZMBGLCNA-N
h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
45357981
Ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoathydrochlorid
Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
23234-43-7
C11H16ClNO3
245.70
MFCD00180750
BQULAXAVRFIAHN-OXZMBGLCNA-N
h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
45357981
Ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoathydrochlorid
Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material D-Tyrosine ethyl ester hydrochloride
Formelmasse 245.7
Reinheit (%) 98%
Menge 5 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'