missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCXR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15188-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
DCXR | |
Polyclonal | |
Unconjugated | |
DCXR | |
Carbonyl reductase II, DCR, DCXR, HCR2, HCRII, KIDCR, Kidney dicarbonyl reductase, L xylulose reductase, P34H, SDR20C1, Sperm surface protein P34H, XR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171408, 51181, 67880 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z4W1, Q91X52, Q920P0 | |
Dcxr | |
DCXR Fusion Protein Ag7272 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |