Learn More
DDX21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10528-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
DDX21 exists as two isoforms, molecular weight of modified isoform one is about 87 -100 kDa, and the post-modified isoform is about 75-85 kDa. 10528-1-AP is a rabbit polyclonal antibody raised against the N-terminal of human DDX21.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an antigen recognized by autoimmune antibodies from a patient with watermelon stomach disease. This protein unwinds double-stranded RNA, folds single-stranded RNA, and may play important roles in ribosomal RNA biogenesis, RNA editing, RNA transport, and general transcription.Spezifikation
DDX21 | |
Polyclonal | |
Unconjugated | |
DDX21 | |
DDX21, DEAD box protein 21, Gu alpha, GUA, GURDB, Nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/GU, RH II/GuA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
317399, 56200, 9188 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3B8Q1, Q9JIK5, Q9NR30 | |
Ddx21 | |
DDX21 Fusion Protein Ag0804 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |